Learn More
Worthington Biochemical Corporation Neonatal Cardiomyocyte Isolation System, 3 kits
Supplier: Worthington Biochemical Corporation LK003303
Kit for performing five separate tissue dissociations each containing up to twelve hearts. Contains single use vials of purified collagenase and trypsin, CMF-HBSS, Leibovitz L-15 media and Falcon cell strainers along with a detailed protocol. The kit is use-tested by Worthington to assure performance. Store at 2-8°C.