Learn More
Applied Biosystems™ NED™ Matrix Standard
Uses the multicomponent matrix to automatically analyze the four different colored fluorescent dye-labeled samples in a single capillary.
Supplier: Applied Biosystems™ 402996
Includes: 1 Tube of NED Matrix Standard
Description
The NED™ Matrix Standard can be used to analyze 6-FAM™, VIC®, NED™ and ROX™-labeled DNA fragments in fragment analysis applications. The standard can be used on the Applied Biosystems® 310 Genetic Analyzers.Order Info
Each
Specifications
Contains 1 tube of NED Matrix Standard. Store at 2-8°C. (Do not freeze). | |
Sequencing | |
NED™ | |
1 Tube |
310 Genetic Analyzer | |
NED | |
Matrix Standard | |
Wet Ice |