Learn More
Thermo Scientific NEBULIZER FOR I CAP Q
Supplier: Thermo Scientific 1317090
PFA-ST MicroFlow Nebulizer. Select a Thermo Scientific™ Nebulizer to meet your laboratory's needs from this range compatible with the Thermo Scientific™ iCAP™ Q ICP-MS analyzer. The standard cyclonic spray chamber on the iCAP Q ICP-MS system accepts all types of nebulizers with a 6mm outside diameter.