Learn More
Cayman Chemical NBQX (sodium salt), 479347-86-9, 50 mg
SDP
Supplier:  Cayman Chemical 1491450
Encompass_Preferred
Molecular Formula C12H6N4O6S • 2Na, Purity ≥98%, Formula Weight 380.2; A water-soluble receptor antagonist which targets both AMPA- and kainate-sensitive glutamate receptors (IC50 = 0.15 and 4.8 M, respectively); does not block the NMDA-sensitive glutamate receptor (IC50 = 90 M).
Catalog No. NC1012646