Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
National Marker™ Write-On Hazardous Communication Labels
For customizing labels
Specifications
Certifications/Compliance | 29 CFR 1910.1200 |
---|---|
Color | Red, Yellow, Blue, White |
Format | Label |
Product Type | Label |
For Use With (Application) | Hazardous materials |
Catalog Number | Mfr. No. | Quantity | Height (English) | Height (Metric) | Width (English) | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Height (English) | Height (Metric) | Width (English) | Price | Quantity & Availability | ||
19-035-033
|
National Marker™
HM42 |
10/Pk. | 6 in. | 15.24 cm | 6 in. | Item Discontinued This item has been discontinued by Fisher Scientific. Please Sign In to view product availability in your area. | N/A | ||
17-984-5A
|
National Marker™
HMS 10P |
5/Pk. | 10 in. | 25.4 cm | 10 in. | Item Discontinued This item has been discontinued by Fisher Scientific. Please Sign In to view product availability in your area. | N/A | ||
17-984-5B
|
National Marker™
HMS 10 R |
1 Ea. | 10 in. | 25.4 cm | 10 in. | Item Discontinued This item has been discontinued by Fisher Scientific. Please Sign In to view product availability in your area. | N/A | ||
17-984-5D
|
National Marker™
HMS 15 R |
1 Ea. | 15 in. | 38.1 cm | 15 in. | Item Discontinued This item has been discontinued by Fisher Scientific. Please Sign In to view product availability in your area. | N/A | ||
17-984-5C
|
National Marker™
HMS 15 P |
5/Pk. | 15 in. | 38.1 cm | 15 in. | Item Discontinued This item has been discontinued by Fisher Scientific. Please Sign In to view product availability in your area. | N/A | ||
Description
- Pressure-sensitive vinyl labels
- Write on and/or use the Personal Protection Numbers and Symbols (Cat. No. 17-981-61D)
- Measure 6 × 6 in. (15.2 × 15.2cm)
Order Info
Available in pack of 10.
Compliance
Comply with 29 CFR1910.1200.
Specifications
29 CFR 1910.1200 | |
Label | |
Hazardous materials |
Red, Yellow, Blue, White | |
Label |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title