Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
National Marker™ Write-On Hazard Warning Labels
Specifications
Format | Label |
---|---|
Material | Paper |
Product Type | Label |
Quantity | 500/Pk. |
Width (English) | 3 in. |
Catalog Number | Mfr. No. | Display Text | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Display Text | Price | Quantity & Availability | |||||
17-986-90C
|
National Marker™
WOL 4 |
NFPA | Item Discontinued This item has been discontinued by Fisher Scientific. Please Sign In to view product availability in your area. | N/A | |||||
17-986-90E
|
National Marker™
WOL 6 |
Biohazard Infectious Waste | Item Discontinued This item has been discontinued by Fisher Scientific. Please Sign In to view product availability in your area. | N/A | |||||
17-986-90B
|
National Marker™
WOL 3 |
Hazard Warning |
Item Discontinued
This item has been discontinued by Fisher Scientific. Please Sign In to view product availability in your area. View alternate product |
N/A | |||||
Description
National Marker Write-On Hazard Warning Labels
Specifications
Label | |
Label | |
3 in. | |
2.54 cm |
Paper | |
500/Pk. | |
1 in. |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title