Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
National Marker™ Paper Shipping Labels for Hazardous and Nonhazardous Waste
Chemical- and weather-resistant labels
Specifications
Certifications/Compliance | EPA and DOT, 17-985-409C or -409D, 17-985-409E |
---|---|
Color | Yellow |
Format | Label |
Material | Paper |
For Use With (Application) | Hazardous waste |
Catalog Number | Mfr. No. | Quantity | Display Text | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Display Text | Price | Quantity & Availability | ||||
17-981-62C
|
National Marker™
HW 10 |
500/Pk. | Hazardous Waste | Item Discontinued This item has been discontinued by Fisher Scientific. Please Sign In to view product availability in your area. | N/A | ||||
17-985-409D
|
National Marker™
HW16 |
500/Pk. | Hazardous Waste (California) | Item Discontinued This item has been discontinued by Fisher Scientific. Please Sign In to view product availability in your area. | N/A | ||||
Description
Order Info
Measure 6 x 6 in.; Pressure-sensitive paper; Roll of 500
Specifications
EPA and DOT, 17-985-409C or -409D, 17-985-409E | |
Label | |
Hazardous waste | |
4 mil | |
6 in. |
Yellow | |
Paper | |
500/Pk. | |
6 in. | |
15.24 cm |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title