Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
National Marker™ Danger: Confined Space Entry Permit Not Required Signs
Specifications
Certifications/Compliance | OSHA 29 CFR 1910.145 |
---|---|
Quantity | 1 Ea. |
For Use With (Application) | Perfect way to alert employees and guests when a hazard is present in your facility |
Displayed Text | DANGER Confined Space Entry Permit not Required |
Product Type | OSHA Safety Sign |
Catalog Number | Mfr. No. | Material | Height (English) | Width (English) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Material | Height (English) | Width (English) | Price | Quantity & Availability | |||
18-999-709
|
National Marker™
D373P |
Vinyl | 7 in. | 10 in. | Item Discontinued This item has been discontinued by Fisher Scientific. Please Sign In to view product availability in your area. | N/A | |||
18-999-708
|
National Marker™
D373RB |
Plastic | 10 in. | 14 in. | Item Discontinued This item has been discontinued by Fisher Scientific. Please Sign In to view product availability in your area. | N/A | |||
Specifications
OSHA 29 CFR 1910.145 | |
Perfect way to alert employees and guests when a hazard is present in your facility | |
OSHA Safety Sign |
1 Ea. | |
DANGER Confined Space Entry Permit not Required |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title