Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
National Marker™ Caution Hazardous Material Storage Area Signs
Alert employees and guests of hazardous areas in a facility
Specifications
Certifications/Compliance | OSHA 29 CFR 1910.196 |
---|---|
Quantity | 1 Ea. |
Displayed Text | CAUTION Hazardous Material Storage Area |
Product Type | Caution Sign |
Color | Black on Yellow |
Catalog Number | Mfr. No. | Material | Dimensions (L x W) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Material | Dimensions (L x W) | Price | Quantity & Availability | ||||
17-982-1J
|
National Marker™
C 310 RB |
Aluminum | 25.4 x 35.6cm (10 x 14 in.) | Item Discontinued This item has been discontinued by Fisher Scientific. Please Sign In to view product availability in your area. | N/A | ||||
17-985-49A
|
National Marker™
C 310 P |
Vinyl | 17.7 x 25.4cm (7 x 10 in.) | Item Discontinued This item has been discontinued by Fisher Scientific. Please Sign In to view product availability in your area. | N/A | ||||
Description
- Yellow and black caution sign
- Available in aluminum and pressure-sensitive vinyl
- Comes in two sizes
1798549A: PS Vinyl; 17.7 x 25.4cm (7 x 10 in.)
179821J: Aluminum; 25.4 x 35.6cm (10 x 14 in.)
Compliance
Comply with OSHA 29 CFR 1910.196
Specifications
OSHA 29 CFR 1910.196 | |
CAUTION Hazardous Material Storage Area | |
Black on Yellow |
1 Ea. | |
Caution Sign |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title