Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
National Marker™ Biohazard: Use Universal Precautions Sign
Sign for hazardous areas advising standard precautions
Specifications
Quantity | 1 Ea. |
---|---|
Height (English) | 7 in. |
Displayed Text | BIOHAZARD Use Universal Precautions |
Product Type | Biohazard Sign |
Catalog Number | Mfr. No. | Width (English) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Width (English) | Price | Quantity & Availability | |||||
18-999-761
|
National Marker™
S95P |
7 in. | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||||
18-999-760
|
National Marker™
S95R |
17 in. | Item Discontinued This item has been discontinued by Fisher Scientific. Please Sign In to view product availability in your area. | N/A | |||||
Description
- Orange sign with black text
- Available in rigid plastic or pressure-sensitive vinyl
- Sized 17.7 × 17.7 cm (7 × 7 in.)
Specifications
1 Ea. | |
7 in. | |
BIOHAZARD Use Universal Precautions | |
Biohazard Sign |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title