Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
National Marker™ Biohazard: Sharps Sign
Sign for entrances of hazardous areas with sharps waste
Supplier: National Marker™ S90R
Description
- Orange sign with black text
- Size 17.7 × 17.7cm (7 × 7 in.)
- Made of plastic
Specifications
Biohazard Sign | |
Plastic | |
7 in. |
Biohazard Sharps | |
1 Ea. | |
7 in. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction