Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
National Marker™ Biohazard: Peligro Biologico Sign
Spanish language biohazard warning sign
Specifications
Quantity | 1 Ea. |
---|---|
Height (English) | 7 in. |
Width (English) | 7 in. |
Displayed Text | BIOHAZARD/PELIGRO BIOLOGICO |
Product Type | Biohazard Sign |
Catalog Number | Mfr. No. | Material | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Material | Price | Quantity & Availability | |||||
19-035-049
|
National Marker™
S91R |
Plastic | Item Discontinued This item has been discontinued by Fisher Scientific. Please Sign In to view product availability in your area. | N/A | |||||
19-035-050
|
National Marker™
S91P |
Vinyl | Item Discontinued This item has been discontinued by Fisher Scientific. Please Sign In to view product availability in your area. | N/A | |||||
Description
- Orange sign with black text
- Sign: 17.7 × 17.7cm (7 × 7 in.)
- Available in plastic or self-sticking polyester
Specifications
1 Ea. | |
7 in. | |
Biohazard Sign |
7 in. | |
BIOHAZARD/PELIGRO BIOLOGICO |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title