Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
National Marker™ Biohazard: Authorized Personnel Only Sign
Sign for hazardous areas are emphasized by graphic
Specifications
Certifications/Compliance | OSHA |
---|---|
Quantity | 1 Ea. |
Height (English) | 7 in. |
Width (English) | 17 in. |
Displayed Text | BIOHAZARD AUTHORIZED PERSONNEL ONLY |
Catalog Number | Mfr. No. | Material | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Material | Price | Quantity & Availability | |||||
18-999-757
|
National Marker™
SA165P |
Vinyl | Item Discontinued This item has been discontinued by Fisher Scientific. Please Sign In to view product availability in your area. | N/A | |||||
18-999-756
|
National Marker™
SA165R |
Plastic | Item Discontinued This item has been discontinued by Fisher Scientific. Please Sign In to view product availability in your area. | N/A | |||||
Description
- Orange sign with black text and graphic
- Available in rigid plastic or pressure-sensitive vinyl
- Size: 17.7 × 43.2cm (7 × 17 in.)
Specifications
OSHA | |
7 in. | |
BIOHAZARD AUTHORIZED PERSONNEL ONLY |
1 Ea. | |
17 in. | |
Safety Sign |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title