Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ NanoDrop™ OneC Microvolume UV-Vis Spectrophotometer PROMO

Maximize your productivity with the Thermo Scientific™ NanoDrop™ OneC Spectrophotometer to quantify and qualify samples before your next experiment.

Supplier:  Thermo Scientific™ 8402742001PR23

Catalog No. 13-400-519PM3


Add to Cart

Description

Description

Our next generation Thermo Scientific™ NanoDrop™ OneC microvolume spectrophotometers are pushing the boundaries once again with the Thermo Scientific™ Acclaro™ Sample Intelligence Software that helps you understand the quality of your sample before you use it in downstream applications, bringing you one step closer to success. NanoDrop™ OneC spectrophotometer features a thermostated, stirred cuvette position for additional life science measurements.

Product Features:

  • Full spectrum microvolume UV-Vis spectrophotometer with cuvette position (stirring, 37°C thermostatting, kinetics)
  • Meas. range 0.4 to 27500 ng/µL dsDNA
  • 190 to 850 nm wavelength meas range
  • Column integrity sensor
  • 2 yr warranty

Quantify and qualify DNA, RNA, and protein samples with only 1 to 2 µL and have full-spectral data in seconds using the Thermo Scientific™ NanoDrop™ OneC Microvolume UV-Vis spectrophotometer.

  • Prevent costly delays — identify contaminants, obtain corrected concentrations, and enjoy success in your downstream applications
  • Collect data fast — Auto-Blank and Auto-Measure modes save time while organizing data
  • Save bench space — user-friendly local control with sleek, standalone design means no PC required

NanoDrop OneC Instrument Highlights

  • Contains both pedestal and cuvette positions to expand experimental flexibility and increase the dynamic range.
  • Measure dilute samples, perform kinetics experiments and take optical density measurements of bacterial cultures.
  • Cuvette position includes temperature control and stirring.

Thermo Scientific™ Acclaro™ Sample Intelligence Software enhances user understanding of sample quality while delivering accurate quantitative measurements. Early identification of contaminants can prevent failure of downstream applications and save days of troubleshooting.

  • Verify sample quality in seconds — identify sample contaminants and obtain corrected concentration results
  • Access information through alerts — receive instant feedback about sample quality with on-demand technical support and guided troubleshooting
  • Rely on accurate results — ensure measurement integrity with embedded sensor and digital image analysis

NanoDrop OneC Enhancements

  • Ergonomic, standalone design — integrated Android™ tablet eliminates the need for a separate computer and provides modern connectivity for seamless data transfer via Wireless*, Ethernet or USB.
  • Auto-Measure and Auto-Blank functions — multi-sample processing can be streamlined with instant measurements that occur when the pedestal arm is down. These functions can be turned ON or OFF with a single tap on the screen.
  • Wider dynamic range — no dilutions required for highly concentrated samples (dsDNA at 27,500 ug/µL) with auto-range pathlength technology.
  • Integrated Learning Center — an archive of answers with technical support documents and educational animations at your fingertips.

NanoDrop One/OneC Spectrophotometer capabilities:

  • Wide spectral range (190 to 850 nm) for measuring a variety of samples types:
    • Peptides (205 nm)
    • DNA and RNA (260 nm)
    • Purified protein (280 nm)
    • Toxicology assays and industrial dyes (490 nm)
    • Gold nanoparticles (520 nm)
    • Colorimetric protein assays (BCA 562 nm, Bradford 595 nm, Modified Lowry 650 nm, Pierce 660 nm)
    • Optical Density measurements (600 nm)
  • Combining the patented** sample-retention system with cuvette capability accommodates low and high concentrations (2.0 to 27,500 ng/ µL dsDNA and 0.06 to 820 mg/mL BSA)
  • Pedestal measurements that require only 0.5 to 2 µL of your sample and do not require sample dilution, even for highly concentrated samples
  • Pre-configured methods for DNA, Protein A280, Microarray, Protein and Labels, Pierce 660, Bradford, BCA, and Lowry
  • User-friendly software that includes custom methods and data export capabilities
  • Instrument comes with a 2-year warranty for worry-free support

* Wi-Fi model not available in certain countries - consult your local NanoDrop distributor
**Patents US6628382 and US6809826
Android is a trademark of Google Inc.

For research use only. Not for diagnostic purposes.
All NanoDrop instruments are approved to CE and UL/CSA.

Specifications

Specifications

NanoDrop™
UV-Vis Spectrophotometer
PR-1 pedestal conditioning solution, PV-1 performance verification solution, USB memory device, Microfiber screen wipe, USA Power Cable
2 Years
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit