Learn More
Thermo Scientific NANODROP EIGHT SPEC PROMO
Manufacturer:  Thermo Scientific 912A11001LD22
This product has been made available for purchase due to recent demand, however information about the product is limited. Please contact customer service with any questions.
Catalog No. 13-400-527PR
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Specifications
Specifications
3% at 0.97 A, 302 nm, 23 ±2°C
NanoDrop™ Eight Spectrophotometer US/CAN
2048-element CMOS Linear Image Sensor
Xenon Flash
190 to 850 nm
1.0 mm, 0.2 mm, 0.1 mm (automatically adjusting)
1 to 2 μL
1 Year
8.2 lb.
0.04 to 200 A (10 mm equivalent)
2.0 to 10,000 ng/μL dsDNA; 0.03 to 145 mg/mL IgG
24 x 33 x 17 cm (9.4 x 13 x 6.7 in.)
100/240 V
<20 sec. (for 8 samples)
3 W at Idle; 15 W at Working Conditions
≤1.8 nm (FWHM at Hg 254 nm)
±1 nm
3.7 kg