Learn More
Polysciences, Inc. Nanobead NIST Traceable Particle Size Standard, 100nm, 15 mL
Supplier: Polysciences, Inc. 6401015
Precision Size Standards are offered in three size groupings: Nanobead, Microbead, and Megabead. They are measured on in-house instruments calibrated with NIST (National Institute of Standards and Technology) Standard Reference Materials. Each product is supplied with a Certificate of Analysis documenting traceability to NIST Standard Reference Materials. The size, standard deviation, coefficient of variation, and lot number are printed on the label for easy reference.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.