Learn More
Daigger & Company NALGENE WASTE CONT & BTL 2L
SDP
Supplier:  Daigger & Company EF21760D
Encompass
Nalgene Benchtop Radioactive Waste Containers are cylindrical shields and lids that securely hold included 2 L polyethylene fluid waste bottle. A lip on the lid prevents slipping. Maximum beta radiation containment is possible with 3/8 inch acrylic. Note: This radioactive waste container includes a 2L wide-mouth HDPE bottle (EF14767A). Replacements for this bottle can be purchased below, within the "Accessories" tab. Key Features •3/8-inch acrylic •Maximum containment •Lip on the lid prevents slipping
Catalog No. NC1020440