Learn More
Thermo Scientific™ Nalgene™ Color-Coded LDPE Wash Bottles
Description
High quality squeeze bottles with color-coded closures for easy identification. Thermo Scientific™ Nalgene™ Color-Coded LDPE Wash Bottles are made from lab quality low density polyethylene (LDPE) for dependably low leachables and chemical compatability with a wide range of lab reagents. Enjoy smooth, precision, sputter-free fluid dispensing with an ergonomic soft squeeze. Ideal for rinsing tubes and lab apparatus, bringing volumetric flasks to final volume and other precision rinsing and solution dispensing needs. The dispensing nozzle is molded in one piece with the bottle closure eliminating risk of separation when the bottle is squeezed. Cut back tip to increase flow.
- Color-coded top allows you to coordinate reagents with industry storage codes.
- Tip can be cut back to provide increased flow.
- Tight fit of draw tube and closure prevents separation when squeezing bottle.
Specifications
Closure Material | Polypropylene |
Closure Size | 28 mm |
Closure Type | Screw Cap |
Diameter (Metric) | 72.9 mm |
Through the Closure Dispensing | Yes |
Product Type | Color-coded Wash Bottle |
Height (Metric) | 224 mm |
Height (English) | 8.8 in. |
Capacity (Metric) | 500 mL |
Capacity (English) | 16 oz. |
Show More |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.