Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

n-Propyl Alcohol, Purified Plus™, ACS Reagent, for organic synthesis, for prep-LC, >99.5%, Honeywell Burdick & Jackson

Catalog Number 6000106
Click to view available options
Quantity:
19 L
204 L
Packaging:
Metal Can
Metal Drum
71-23-8
C3H8O
60.10
MFCD00002941
BDERNNFJNOPAEC-UHFFFAOYSA-N
1-propanol, propanol, propyl alcohol, n-propanol, n-propyl alcohol, ethylcarbinol, 1-hydroxypropane, optal, osmosol extra, propylic alcohol
1031
CHEBI:28831
propan-1-ol
CCCO
71-23-8
C3H8O
60.10
MFCD00002941
BDERNNFJNOPAEC-UHFFFAOYSA-N
1-propanol, propanol, propyl alcohol, n-propanol, n-propyl alcohol, ethylcarbinol, 1-hydroxypropane, optal, osmosol extra, propylic alcohol
1031
CHEBI:28831
propan-1-ol
CCCO
CAS Max % 100.0000%
UN Number UN1274
Beilstein 1098242
Solubility Information Miscible in Water at 20°C
Formula Weight 60.09
Percent Purity >99.5%
Grade ACS Reagent
Density 0.8053 g/cm3
Boiling Point 97.2°C
Quantity 204 L
Flash Point 23°C
Packaging Metal Drum
Water 0.1% max.
Assay by GC Analysis
Chemical Name or Material n-Propyl Alcohol, Purified Plus™, ACS Reagent, for organic synthesis, for prep-LC, >99.5%
Show More Show Less
ShelfLife 547 days from date of manufacture
DOTInformation Transport Hazard Class: 3; Packing Group:3: III; Proper Shipping Name: n-Propanol
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit