Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
n-Propanol (Peroxide-Free/Sequencing), Fisher BioReagents
Click to view available options
Quantity:
500 mL
Packaging:
Glass Bottle
Description
n-Propanol is used for extracting ethidium bromide from DNA samples.
Specifications
pH | 7 |
Melting Point | -127°C |
Color | Colorless |
Acidity | 0.02% max. |
Density | 0.800g/cm³ |
Boiling Point | 97°C |
Quantity | 500 mL |
Identification | Pass Test |
Packaging | Glass Bottle |
Physical Form | Liquid |
Show More |
Safety and Handling
Recommended Storage | RT |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction