Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
N,N-Dimethylformamide, For gas chromatography ECD and FID SupraSolv™, MilliporeSigma™
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Glass Bottle
Specifications
pH | 7 (at 200g/L 20°C) |
Melting Point | -61°C |
Color | Colorless |
Density | 0.94g/cm3 at 20°C |
Boiling Point | 153°C |
Quantity | 2.5 L |
Flash Point | 58°C (136°F) |
Ignition Point | 410°C (770°F) |
Odor | Weakly amine-like |
Packaging | Glass Bottle |
Show More |
Safety and Handling
Hazard Category | H360D May damage the unborn child. |
Hazard Statement | H226 Flammable liquid and vapor. |
Precautionary Statement | H312 + H332 Harmful in contact with skin or if inhaled. |
Recommended Storage | 5°C to 30°C (41°F to 86°F); Keep container tightly closed in a dry and well-ventilated place. Keep away from heat and sources of ignition. Keep locked up or in an area accessible only to qualified or authorized persons. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction