Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
N,N-Dimethylformamide, BioSyn™, for biosynthesis and applications requiring low water content, >99.9%, Burdick & Jackson, Honeywell
Click to view available options
Quantity:
4 x 4 L
Description
- Ultra-low water specifications
- Low amine specifications
- Low particulate levels
- High solvent purity
Specifications
CAS Max % | 100.00 |
Density | 0.949 g/cm3 |
Boiling Point | 153°C |
Quantity | 4 x 4 L |
Flash Point | 58°C |
Packaging | Glass Bottle |
Physical Form | Liquid |
Grade | Low Water |
Safety and Handling
Hazard Category | Danger |
Hazard Statement | H360+H332+H319+H226 |
Precautionary Statement | P280+P271+P264+P261+P243+P242+P241+P240+P233+P210+P202+P201 |
Recommended Storage | Room Temperature |
WARNING: Cancer - www.P65Warnings.ca.gov
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction