Learn More
Mystery of Lyle and Louise: An Inky Lead™ Questioned Documents Analysis Forensic Science Curriculum

Introduce methods of handwriting and ink analysis.
$117.20 - $164.30
Specifications
Certifications/Compliance | Meets National Science Education Standards |
---|---|
Grade | 9 to Undergraduate |
Includes | 12 tlc plates, six bottles of solvent, six development chambers, six protractors, box of microcaps, forged receipts, copies of forged receipts, teaching manual |
Class Size | 30 Students |
Includes: 12 thin-layer chromatography (TLC) plates, six bottles of solvent, six development chambers, six protractors, box of microcaps, forged receipts, copies of forged receipts, teaching manual
Catalog Number | Mfr. No. | For Use With (Application) | Product Type | Requires | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | For Use With (Application) | Product Type | Requires | Price | Quantity & Availability | |||
S94584
![]() |
Crosscutting Concepts LLC
VXH10132 |
For introduce methods of handwriting and ink analysis | Mystery Of Lyle And Louise: An Inky Lead™ Questioned Documents Analysis Forensic Science Curriculum Kit | Inky Lead™ questioned documents analysis kit, scissors, 12 glass microscope slides, 6 to 12 forceps, lab gloves |
Each for $164.30
|
|
S94584R
![]() |
Crosscutting Concepts LLC
VXH10278 |
For used to replenish consumable components | Mystery Of Lyle And Louise: An Inky Lead™ Questioned Documents Analysis Forensic Science Refill Kit | An inky lead™ questioned documents analysis kit, scissors, glass microscope slides, forceps, lab gloves |
Each for $117.20
|
|
Description
After learning about handwriting analysis and questioned document examination, students set out to determine if any receipts from Tumbling Water Land Development Company have been forged. To do this, students must determine objective criteria for handwriting analysis, and then apply these criteria when examining suspect receipts.
- Analyze handwriting
- Dissolve the ink from numbers on the suspect invoices
- Use thin-layer chromatography to create an ink profile for each of the samples taken from the receipt
- Determine if the same ink was used to fill out the entire form
- Develop a complete theory of the suspected forgery
- Class size: six groups of up to five students per group
- For high school and undergraduate courses
- Meets National Science Education Standards
Specifications
Meets National Science Education Standards | |
9 to Undergraduate | |
12 tlc plates, six bottles of solvent, six development chambers, six protractors, box of microcaps, forged receipts, copies of forged receipts, teaching manual | |
30 Students |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.