Learn More
Mystaire™ Aura™ Elite Series Ductless Fume Hood Pre-Filter
For Aura Elite Series 30, 42 and 54 in. Ductless Fume Hoods. Protects the adsorption filter from submicron particulate.
Supplier: Mystaire™ MYPRE24
Description
- Electrostatically charged fiber filter
- Designed to protect adsorption filter from contaminants such as smoke, acid mists, and other submicron particulates
- All units are initially supplied with a pre-filter
Specifications
Particulate Prefilter |