Learn More
Mystaire™ Evidence Drying Cabinet FR Series Main Filter
For use with the Mystaire™ FR-3100 and FR-6100 Evidence Drying Cabinet. Mystaire™ filters are manufactured to the strictest quality standards in the industry to ensure their capability to meet your application challenges.
Supplier: Mystaire™ FE1019
Description
- Made in the U.S.A.
- Each filter application is reviewed to ensure a proper filtration package is selected
- Filter performance verified by Fourier Transform Infrared Spectroscopy (FTIR)
Specifications
FR-3100 and FR-6100 Evidence Drying Cabinet | |
Filter | |
Gas Phase Carbon | |
Odors of Decomposition |