Learn More
Lonza Walkersville MycoZap Plus-PR
Supplier: Lonza Walkersville VZA2021
MycoZap Plus-PR is a ready-to-use solution of innovative antibiotics for protecting cell cultures from a broad range of common contaminants. The special formulation of MycoZap Plus-PR allows a reliable and gentle protection of primary cells which are usually delicate in means of chemical robustness and usually do not tolerate high antibiotic concentrations to prevent microbial contamination. MycoZap Plus-PR is active against mycoplasma, but also Gram(-) and Gram(+) bacteria as well as yeast and fungi. MycoZap Plus-PR can be used continuously and replace the old-fashioned Pen/Strep formulation.Benefits:Active against mycoplasma, Gram(-) and Gram(+) bacteria as well as yeast and fungiComplete solution replacing Pen/Strep formulationApplications:MycoZap Plus-PR optimized for gentle protection of primary cellsContent:10 x 1 mL MycoZap Plus-PR Antibiotic
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.