Learn More
Bionique Testing Labs Inc Mycoplasma Control Slides, 5/pk
Supplier: Bionique Testing Labs Inc M600/DUP
Five (5) Control Slides are provided with each order for in-house DNA fluorochrome staining. Each slide contains two negative and two positive mycoplasma controls, fixed and ready for staining. These Mycoplasma Quality Control slides are designed to verify the quality of reagents and staining method used in the technique of DNA fluorochrome staining for the detection of mycoplasma contamination.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.