Learn More
Lonza Walkersville MycoAlert PLUS Assay Buffer
Supplier: Lonza Walkersville LT27292
The MycoAlert PLUS Mycoplasma Detection Kit can be used to test fresh media or supplements for mycoplasma contamination. A specific protocol is recommended for testing neat samples outlined in the ‘Instructions for Use’. The protocol involves a sample dilution step that helps eliminating any inhibitory influence of the culture media components to the MycoAlert PLUS Assay Reactions. The protocol includes a sample dilution step of 1:10 with the MycoAlert PLUS Assay Buffer.Content:1 x 20 mL MycoAlert PLUS buffer
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.