Learn More
Lonza Walkersville MycoAlert PLUS Mycoplasma Detection Kit (100 Tests)
Supplier: Lonza Walkersville LT07710
Lonza’s MycoAlert Mycoplasma Detection Assays are selective biochemical tests to detect Mycoplasma contamination in cell cultures. They exploit the activity of mycoplasmal enzymes which are found in the vast majority of about 200 mycoplasma species, but are not present in eukaryotic cells.Benefits:Easy assay: Simply add two reagents to the culture supernatant and perform two luminescence readingsResults in < 20 minEasy interpretation of resultsApplications:Detects all common mollicute contaminations (Mycoplasma, Acholeplasma, Entomoplasma and Spiroplasma; except Ureaplasma) Suited for tube luminometers, plate luminometers and multifunctional readersContent:1 x 10 mL MycoAlert PLUS Reagent (lyophilized)1 x 10 mL MycoAlert PLUS Substrate (lyophilized)1 x 20 mL MycoAlert PLUS Assay Buffer
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.