Learn More
Cliawaived Inc MultiTox Urine Control (Negative) – 25mL
Supplier: Cliawaived Inc MULTITOXNC25E
Multi-Tox Urine Control (Negative )- 25mL. FDA Cleared - Negative control for use with all point of care urine drug test cups and cards.
This item is not returnable.
View return policy