Learn More
Cliawaived Inc Multi-Tox Combo Urine Control Set (2x 25mL, 1 Pos & 1 Neg)
SDP
Supplier:  Cliawaived Inc MULTITOXPC25+NC25
Encompass
Multi-Tox Combo Urine Control Set (2x 25mL) includes 1 Pos & 1 Neg. Positive Controls for use with all point of care cups and cards that include THC,COC, PCP, MOR or OPI, AMP, mAMP, BAR, BZO, MTD, MTQ, PPX, TCA, MDMA, OXY, BUP G, and EDDP.
Catalog No. NC1494155
Due to product restrictions, please to purchase or view availability for this product.
missing translation for 'nonReturnableMessage' missing translation for 'returnPolicyLabel'
missing translation for 'provideContentCorrection'
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
missing translation for 'productTitle'