Learn More
Andwin Scientific MULTISTAGE (DUAL-STAGE) GAS
SDP
Supplier:  Andwin Scientific 55850422
Encompass_Preferred
Multistage Gas Regulator Argon/nitrogen/helium. Brass body. Stainless steel diaphragms. PTFE encapsulated seat. 6.35mm NPT valve and body outlets. Inlet CGA: 580. Flow Capacity: 765/915/2430CFH. Delivery range: 1-50psi.
Catalog No. NC0393866