Learn More
Gilson™ Multichannel Pipetman™ L Pipetter Trade-in Promotion
Designed for performance and comfort, Pipetman™ L Multichannel Pipette comes in a comprehensive range of models, adapted to your needs in the lab. Comfortable, precise, accurate, and reliable, Pipetman™ L variable volume multichannel models offer optimized performance across channels for uniform sample delivery and are perfect solutions when pipetting on microplates.
Specifications
Compatible Tips | Diamond™ D200 Tips |
---|---|
Adjustable Tip Spacing | No |
Autoclavable | Autoclavable |
Color | Dark Blue |
Dispensing Speed | Manual |
Catalog Number | Mfr. No. | Model | Number of Channels | Volume (Metric) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Model | Number of Channels | Volume (Metric) | Price | Quantity & Availability | |||
FA10035TG
|
Gilson™
FA10035T |
P8X200L | 8 | 20 to 200 μL | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
FA10036TG
|
Gilson™
FA10036T |
P12X200L | 12 | 20 to 200 μL | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
FA10037TG
|
Gilson™
FA10037T |
P8x300L | 8 | 20 to 300 μL | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
FA10038TG
|
Gilson™
FA10038T |
P12x300L | 12 | 20 to 300 μL | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
Description
Available as a fixed or variable volume, Pipetman™ L air-displacement pipettes cover a full volume range, including:
• Variable volume single channel models covering a volume range from 0.2 μL to 10 mL
• Variable volume multichannel models covering a volume range from 0.5 μL to 1200 μL, including four models equipped with V-ring (VR) at the bottom of the tip holders for a proper fit with most tip brands, covering a volume range from 20 µL to 300 µL
• Fixed volume single channel models covering a volume range from 1 μL to 5000 μL
Specifications
Diamond™ D200 Tips | |
Autoclavable | |
Manual | |
Pipette Tips | |
Polypropylene | |
Non-sterile | |
Certified Quality |
No | |
Dark Blue | |
No | |
Ergonomic | |
Pipetman™ L | |
Multichannel Pipetter |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.