Learn More
MSA™ Safe Escape CBRN Escape Hood
Supplier:  MSA™ 10047117
Catalog No. 19-166-670
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Description
Description
Guards against HD vapor, GB, Ammonia, Cyanogen chloride, Cyclohexane, Formaldehyde, Hydrogen cyanide, Hydrogen sulfide, Nitrogen dioxide, Phosgene, Phosphine, sulfur dioxide
Certifications
NIOSH - 10052317
Specifications
Specifications
Escape Hood
High-performance laminate material
Product Suggestions
Product Suggestions