Learn More
MSA™ Quick-Fill™ Kit, Portaire Portable Air-Supply System
Portable air supply source allows for rapid transfer of air from a high-pressure source into a cylinder containing less pressure..
Supplier: MSA™ 807053
Description
- Recharge both low and high pressure operating systems
- Quick connect design
- Rapid fill rates between 40 and 60 seconds
- Approved for use in IDLH environments
Specifications
807053 | |
Quick Fill Kit |