Learn More
MSA™ Custom Air V™ Escape Respirator
Provides emergency air in highly toxic or deadly environments
Supplier:  MSA™ 484353
 View more versions of this product
Catalog No. 18-999-3114
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Description
Description
Five minute high-flow cylinder meets stringent airflow requirements for extremely aerobic escape applications
Quick-retracting protective cover saves precious time when donning the hood
Choice of lower cost aluminum or lighter weight composite cylinder
Choice of carbon fiber or aluminum cylinder
Five minute high flow rate of 72lpm
Easy egress from tightly-spaced areas
Can withstand exposure to temperatures from -17.8° to +71.1°C (0° to 160°F)
Urethane hood for industrial operations
  • Air flows directly on to the lens to minimize fogging
  • Hood includes a muffler for quieter operation
Color coded carriers
  • Made of highly visible color-coded nylon
  • Cylinder harness is made of flexible, abrasion-resistant nylon
Compliance
NIOSH: TC-13F-217
Specifications
Specifications
NIOSH
2216 psig
72 L/min.
Lighter weight compressed-air cylinder, urethane hood with muffler, color coded nylon carriers
-17.8° to 71.1°C
Escape Respirator
5 min.
Escape toxic gasses, vapors or oxygen deficiency
0° to 160°F
Product Suggestions
Product Suggestions