Learn More
MSA™ Comfo Classic Probed Respirator Facepieces
$130.60 - $135.70
Specifications
Product Type | Probed Respirator Facepiece |
---|---|
For Use With (Application) | Asbestos abatement, painting, sanding, grinding, welding |
Includes | Built-in probe assembly, solid brass probe and brass fitting |
Material | Silicone |
Catalog Number | Mfr. No. | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Size | Price | Quantity & Availability | |||||
18-999-3676
|
MSA™
808258 |
Small |
Each for $130.60
|
|
|||||
18-999-3674
|
MSA™
808252 |
Medium |
Each for $135.70
|
|
|||||
18-999-3675
|
MSA™
808264 |
Large |
Each for $135.70
|
|
|||||
Specifications
Probed Respirator Facepiece | |
Asbestos abatement, painting, sanding, grinding, welding | |
Built-in probe assembly, solid brass probe and brass fitting | |
Silicone |