Learn More
MSA™ Airline Particulate Filter
Particulate filter for use with MSA air distribution systems.
Supplier:  MSA™ 76367
Catalog No. 19-815-654
Add to Cart
Description
Description
  • Particulate filter
  • For use with air distribution systems
Specifications
Specifications
95% at 0.3μ
Filter, receptacle, washer, nut and insert
76367
Particulate Filter