Learn More
MQuant™ Non-Bleeding pH Indicator Strips
SCI_ED
Since the indicator does not bleed, the strip can be left in the measurement medium without contaminating it.
Supplier:  MilliporeSigma 1.09532.0001/EMD
 View more versions of this product
Catalog No. S07768
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Description
Description
  • Biologically degradable polyester foil
  • Contain special indicator dyes covalently bound to reagent papers
  • Rapid on-the-spot determination
  • Tests checked and calibrated using certified buffer solutions
  • Buffer solutions are traceable to primary reference materials from NIST™ and PTB
  • Suited for all media in environmental analysis and in industrial in-process controls
Specifications
Specifications
7.5 to 14
14
10°C to 25°C
Environmental analysis and industrial in-process controls, calibration and checking of the products by certified buffer Solutions, rapid on-the-spot pH determination, non-bleeding even in strong alkaline solutions for accurate pH determination
100 Pack
Acilit pH Indicator Strip
Product Suggestions
Product Suggestions
Safety and Handling
Safety and Handling
ShelfLife : 3 to 5 Years