Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MP Biomedicals™ RNaid™ Kit
RNaid™ Kit provides a simple and convenient way to isolate total RNA from agarose and polyacrylamide gels and from solutions.
$370.00 - $394.00
Specifications
Sample Type | Enzymatic Reactions, Solutions, Agarose Gels, Polyacrylamide Gels |
---|---|
For Use With (Application) | To isolate, purify RNA (75 bases to 10 kb) from native, denaturing agarose/polyacrylamide gels |
Quantity | 200 Preps |
Catalog Number | Mfr. No. | Product Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Product Type | Price | Quantity & Availability | |||||
MP111007200
|
MP Biomedicals™
111007200 |
RNAMATRIX™ Slurry |
Each for $370.00
|
|
|||||
MP111107200
|
MP Biomedicals™
111107200 |
RNAMATRIX™ Slurry plus SPIN Filter |
Each for $394.00
|
|
|||||
Description
- Kit is designed to isolate and purify RNA (75 bases to 10 kb) from native and denaturing agarose and polyacrylamide gels.
- Removal of unincorporated nucleotides, buffer salts, and other reagents from enzyme reactions.
- The purified RNA is ready for reverse transcription and PCR, labeling and microarray analysis, and other common applications.
Specifications
Enzymatic Reactions, Solutions, Agarose Gels, Polyacrylamide Gels | |
To isolate, purify RNA (75 bases to 10 kb) from native, denaturing agarose/polyacrylamide gels | |
200 Preps |
Unless specified otherwise, for research or further manufacturing use only, not for direct human use.
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title