Learn More
MP Biomedicals™ Protein A from Staphylococcus aureus, 9–11mg
A highly stable cell surface receptor produced by several strains of Staphylococcus aureus which can conjugate with various reporter molecules without affecting the antibody binding site on the molecule
$83.00 - $957.00
Specifications
Product Type | Protein A from S. aureus |
---|---|
Form | Lyophilized |
For Use With (Application) | ELISA, Immunohistochemistry, Western Blotting |
Content And Storage | -20°C |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
ICN15005101
|
MP Biomedicals™
0215005101 |
1 mg |
Each for $83.00
|
|
|||||
ICN15005105
|
MP Biomedicals™
0215005105 |
5 mg |
Each for $268.00
|
|
|||||
ICN15005120
|
MP Biomedicals™
0215005120 |
20 mg |
Each for $957.00
|
|
|||||
Description
Consists of a single polypeptide chain of molecular weight 42 kDa, containing four repetitive domains rich in aspartic and glutamic acids but devoid of cysteine. Contains little or no carbohydrate and only 4 tyrosine residues and no tryptophans.
- Gene Id: Staphylococcus aureus subsp. aureus NCTC 8325 …SAOUHSC_00069(3919448)
- Binding Capacity: 9–11mg of human IgG per mg solid (Lit.)
- Extinction Coefficient: E1%280 = 1.4 (Lit.)
- Presentation: White frozen solid
- Isoelectric Point: 4.85 to 5.10(Lit.)
Specifications
Protein A from S. aureus | |
Lyophilized | |
ELISA, Immunohistochemistry, Western Blotting | |
-20°C |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.