Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MP Biomedicals™ Protein A, Fc Binding Grade
Fc Binding Grade, White Lyophilized Powder. Routinely used for immunoprecipitation assays.
$87.00 - $957.00
Specifications
Binding Capacity | 12 to 15 mg of Human lgG/mg solid |
---|---|
For Use With (Application) | For detection of immunglobulins, purification |
Content And Storage | -20°C |
Form | Lyophilized Powder |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
ICN15389101
|
MP Biomedicals™
0215389101 |
1 mg |
Each for $87.00
|
|
|||||
ICN15389105
|
MP Biomedicals™
0215389105 |
5 mg |
Each for $263.00
|
|
|||||
ICN15389120
|
MP Biomedicals™
0215389120 |
20 mg |
Each for $957.00
|
|
|||||
Description
- Binding capacity 12-15mg of human lgG/mg solid
- Carboxy truncated material containing 301 amino acid residues
- White lyophilized powder
- Source: Staphlococcus aureus , Cowan strain-1
Specifications
12 to 15 mg of Human lgG/mg solid | |
For detection of immunglobulins, purification | |
-20°C | |
Lyophilized Powder |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title