Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MP Biomedicals™ ImmunO™ Immunodiffusion Plates
Gel filled, Uncut
Supplier: MP Biomedicals™ 08642791
Description
- Each plate contains 1.2% agarose (buffer-PBS plus 0.1% Na Azide and 0.01% Merthiolate).
- The high quality immunodiffusion grade agarose and 5% PEG greatly enhance the sensitivity or the immunodiffusion assay
- Store at 2 to 8°C
Specifications
Polystyrene | |
Immunodiffusion Plate | |
2 to 8°C |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.