Learn More
MP Biomedicals™ FastDNA™-96 Soil Microbe DNA Extraction Kit

FastDNA™-96 Soil Microbe DNA Extraction Kit is designed to quickly and efficiently isolate humic-free, PCR-ready genomic DNA from a variety of soil samples.
Supplier: MP Biomedicals™ 119696200
Description
- Product Overview: The FastDNA-96™ Soil Microbe DNA Kit quickly and efficiently isolates humic-free, PCR-ready genomic DNA from soil samples in approximately 50 minutes. Gram(+/-) bacteria, fungi, plant and animal tissues, algae, spores and many other members of a soil population including clay, sandy, silty, peaty, chalky, and loamy soils, are efficiently lysed in approximately 60 seconds with the FastPrep-96™ Instrument.
- Sample Type: Feces, Soil, Wastewater, Environmental Water, Sludge
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Kit | |
2 x 96 Preps | |
1 Ea. |
Feces, Soil, Wastewater, Environmental Water, Sludge | |
FastDNA-96 Kits |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.