Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse IgG1 Negative Control, Mouse, Clone: Ci4, Isotype Control, MilliporeSigma™
Mouse Isotype Control
Supplier: MilliporeSigma MABC002
Description
Mouse IgG1 Negative Control Monoclonal Antibody validated for use in Flow Cytometry and Immunofluorescence.Specifications
IgG1 | |
Monoclonal | |
0.5mg/mL | |
Protein A Purified mouse immunoglobulin in phosphate buffered saline, pH 7.4, containing 0.2% bovine Serum albumin and 0.1% sodium azide. | |
Chicken immunoglobulin | |
500 μL | |
Chicken | |
Purified |
Flow Cytometry, Immunofluorescence | |
Ci4 | |
Unconjugated | |
Mouse | |
Protein A purfied | |
Primary | |
2-8°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles. | |
IgG1 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction