Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse anti-Rat IgG2c, Clone: A92-3, Secondary Antibody, BD
Supplier: BD Biosciences 553910
Description
The A92-3 antibody reacts specifically with rat IgG2c. It does not react with other Ig isotypes.
Host Species: Mouse
Clone: A92-3
Isotype: IgG1, κ
Species Reactivity: Rat
Immunogen: Pooled rat IgG2c
ELISA
Specifications
Mouse anti-Rat IgG2c | |
A92-3 | |
Mouse | |
0.5 mg | |
Immunology | |
Rat |
Monoclonal | |
0.5mg/mL | |
Affinity Purified | |
RUO | |
Secondary | |
IgG1 κ |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction