Learn More
Mott Manufacturing Wall Cabinet Accessory, Toe Space Filler

Mott toe space filler is used to close gaps once base cabinet has been installed.
$36.65
Catalog Number | Mfr. No. | Location | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Location | Price | Quantity & Availability | |||||
04-000-012
![]() |
Mott Manufacturing
9900001NE |
Zone 4 |
Each for $36.65
|
|
|||||
04-000-099
![]() |
Mott Manufacturing
9900001MGC |
Zone 7 |
Each for $36.65
|
|
|||||
04-000-534
|
Mott Manufacturing
9900001SB |
Zone 10 |
Each for $36.65
|
|
|||||
04-000-795
|
Mott Manufacturing
9900001CC |
Zone 1 |
Each for $36.65
|
|
|||||
04-000-186
|
Mott Manufacturing
9900001DT |
Zone 5 |
Each for $36.65
|
|
|||||
Description
- For use with Mott Wall Cabinets
- Used for corner applications
- Zone 1 (Use CC suffix): Montana (Eastern), Minnesota, North Dakota, South Dakota, Wyoming
- Zone 2 (Use WJ suffix): Iowa, Kansas, Missouri, Nebraska, Wisconsin
- Zone 3 (Use SL suffix): Alabama, Florida, Georgia, Illinois, Kentucky, Tennessee
- Zone 4 (Use NE suffix): Connecticut, Delaware, District of Columbia, Maine, Maryland, Massachusetts, New Hampshire, North Carolina, Rhode Island, South Carolina, Vermont, Virginia, West Virginia
- Zone 5 (Previous DT suffix): Moved to Zone 9
- Zone 6 (Previous CF suffix): Moved to Zone 9
- Zone 7 (Use MGC suffix): Arizona, Arkansas, Colorado, Louisiana, Mississippi, New Mexico, Oklahoma, Texas
- Zone 8 (Use S suffix): New Jersey, New York, Pennsylvania (Eastern)
- Zone 9 (Use SV suffix): Michigan, New York State (Upper), Ohio, Pennsylvania (Western)
- Zone 10 (Use SB suffix): Alaska, California, Hawaii, Idaho, Montana (Western), Nevada, Oregon, Utah, Washington
Order Info
When ordering, be sure to select items with the letter code suffix that matches the state that items are to be DELIVERED to
Specifications
Space Filler |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.