Learn More
Motic™ Moticam S3 and Analysis Basic Software
Moticam S3 delivers a mid-size resolution of 3 MP, keeping the balance between speed and resolution. A universal tool for teaching and routine lab, for biology, medicine and industry, the sCMOS sensor guarantees state of the art imaging results for an unbeatable price.
Supplier: Motic™ MBS3
Description
Motic™'s new C-mount ready sCMOS cameras are the newest members of the Moticam family of professionally designed and engineered cameras. It is easy enough for the microscopy novice, but sophisticated enough for the professional. With back illuminated sensor technology and speedy data transfer, this camera creates superbly detailed images and smooth video captures.
- Included Motic™ Images Plus 3.0 for PC and Mac software enables live and captured images at a maximum quality
- Edit, measure, and share images or reports with its comprehensive software
- 3 MP sCMOS camera with USB 3.1 (2048 x 1536 pixels at 60 fps or 1920 x 1080 pixels at 60 fps)
- Requires: 2 GHz multi-core processor or above, USB 2.0 or USB 3.0 (for high speed transmission), 8 GB RAM, 5 GB (Free Space) hard disk, 1 GB Video RAM on video card
Specifications
Microscope Camera | |
CS Ring Adaptor, USB 3.1 Cable, Motic 4-dot Calibration Slide, Motic Images Plus 3.0 for PC/OSX/Linux | |
1/2.8 in. |
Moticam S | |
sCMOS |