Learn More
Motic™ Moticam S1 and Analysis Basic Software
The Moticam S1 is ideal for back-illuminated sensor technology and speedy data transfer (high frame rates up to 120 fps). It creates an optimal imaging environment and makes capturing images easy.
Supplier: Motic™ MBS1
Description
Motic™'s new C-mount ready sCMOS cameras are the newest members of the Moticam family of professionally designed and engineered cameras. It is easy enough for the microscopy novice, but sophisticated enough for the professional. With back illuminated sensor technology and speedy data transfer, this camera creates superbly detailed images and smooth video captures.
- Included Motic™ Images Plus 3.0 for PC and Mac software enables live and captured images at a maximum quality
- Edit, measure, and share images or reports with its comprehensive software
- 1.2 MP sCMOS camera with USB 3.1 (1280 x 960 pixels at 120 fps or 640 x 480 at 240 fps)
- Requires: 2 GHz multi-core processor or above, USB 2.0 or USB 3.0 (for high speed transmission), 8 GB RAM, 5 GB (Free Space) hard disk, 1 GB Video RAM on video card
Specifications
Microscope Camera | |
CS Ring Adaptor, USB 3.1 Cable, Motic 4-dot Calibration Slide, Motic Images Plus 3.0 for PC/OSX/Linux | |
1/3 in. |
Moticam S | |
sCMOS |