Learn More
Motic™ Moticam PROS5 Plus and Analysis Basic Software
Moticam Pro S5 Plus presents a 2/3 in. sensor with 5 MP at a maximized frame rate of up to 70 fps in full resolution. The peak of Motic™'s Pro Series, this camera covers all important aspects of a professional imaging solution.
Supplier: Motic™ 1100600101121
Description
Motic™'s new C-mount ready sCMOS cameras are the newest members of the Moticam family of professionally designed and engineered cameras. It is easy enough for the microscopy novice, but sophisticated enough for the professional. With back illuminated sensor technology and speedy data transfer, this camera creates superbly detailed images and smooth video captures.
- Included Motic™ Images Plus 3.0 for PC and Mac software enables live and captured images at a maximum quality
- Edit, measure, and share images or reports with its comprehensive software
- 5 MP sCMOS camera with USB 3.1 (2448 x 2048 pixels at 68.3 fps or 1224 x 1024 pixels at 175.8 fps)
- Requires: 2 GHz multi-core processor or above, USB 2.0 or USB 3.0 (for high speed transmission), 8 GB RAM, 5 GB (Free Space) hard disk, 1 GB Video RAM on video card
Specifications
Microscope Camera | |
CS Ring Adaptor, USB 3.1 Cable, Motic 4-dot Calibration Slide, Motic Images Plus 3.0 for PC/OSX/Linux | |
2/3 in. |
Moticam S | |
sCMOS |